Tisk…

2-Bromo-4-chloro-6-nitroaniline

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Bromo-4-chloro-6-nitrophenylamine

OR7021-5G OR7021-100G OR7021-25G
APOSOR7021-5GEA 622 CZK
APOSOR7021-5G APOSOR7021-100G APOSOR7021-25G
2-Bromo-4-chloro-6-nitroaniline
2-Bromo-4-chloro-6-nitroaniline
Vzorec: C₆H₄BrClN₂O₂
MW: 251.4700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08275709
CAS číslo: 827-25-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR