Tisk…

5-Phenyl-3-isoxazolecarboxylic acid

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 5-Phenylisoxazole-3-carboxylic acid

OR70203-1G OR70203-10G OR70203-25G
APOSOR70203-1GEA 2070 CZK
APOSOR70203-1G APOSOR70203-10G APOSOR70203-25G
5-Phenyl-3-isoxazolecarboxylic acid
5-Phenyl-3-isoxazolecarboxylic acid
Vzorec: C₁₀H₇NO₃
MW: 189,1700 g/mol
Bod tání: 160…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00464221
CAS číslo: 14441-90-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR