4-Aminobutyraldehyde diethyl acetal
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4,4-Diethoxybutan-1-amine, 4-Aminobutanal diethyl acetal
OR70118-100G
OR70118-25G
OR70118-5G
APOSOR70118-100GEA
6750
CZK
APOSOR70118-100G
APOSOR70118-25G
APOSOR70118-5G
4-Aminobutyraldehyde diethyl acetal
4-Aminobutyraldehyde diethyl acetal
Vzorec:
C₈H₁₉NO₂ Bod varu: 196 °C Hustota: 0,933 g/cm³ (25 °C) Bod vzplanutí: 62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008227 CAS číslo: 6346-09-4 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...