3-Chloro-5-methoxybenzaldehyde 95+%
Dodavatel: Apollo Scientific
|
Varování
|
OR70077-10G
OR70077-1G
OR70077-5G
OR70077-25G
APOSOR70077-10GEA
7050
CZK
APOSOR70077-10G
APOSOR70077-1G
APOSOR70077-5G
APOSOR70077-25G
3-Chloro-5-methoxybenzaldehyde 95+%
3-Chloro-5-methoxybenzaldehyde
Vzorec:
C₈H₇ClO₂ MW: 170,5900 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08234653 CAS číslo: 164650-68-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...