Morpholine-3,5-dione
Dodavatel: Apollo Scientific
OR70072-1G
OR70072-5G
APOSOR70072-1GEA
9460
CZK
APOSOR70072-1G
APOSOR70072-5G
Morpholine-3,5-dione
Morpholine-3,5-dione
Vzorec:
C₄H₅NO₃ Bod tání: 142…145 °C Storage Temperature: Ambient |
MDL Number:
MFCD01863549 CAS číslo: 4430-05-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...