4-Bromobenzaldehyde dimethyl acetal
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-Bromo-4-(dimethoxymethyl)benzene, 4-Bromo-α,α-dimethoxytoluene
OR70059-100G
OR70059-25G
APOSOR70059-100GEA
3610
CZK
APOSOR70059-100G
APOSOR70059-25G
4-Bromobenzaldehyde dimethyl acetal
4-Bromobenzaldehyde dimethyl acetal
Vzorec:
C₉H₁₁BrO₂ MW: 231,09 g/mol Bod varu: 254 °C Hustota: 1,383 g/cm³ (25 °C) Bod vzplanutí: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD01320499 CAS číslo: 24856-58-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...