Tisk…

2-(tert-Butoxycarbonylamino)-1-ethanol

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: N-(2-Hydroxyethyl)carbamic Acid tert-Butyl Ester, 2-(tert-Butoxycarbonyl)ethanolamine, N-(tert-Butoxycarbonyl)ethanolamine, 2-Boc-ethanolamine, tert-Butyl N-(2-Hydroxyethyl)carbamate, 2-(Boc-amino)-1-ethanol, Boc-Glycinol

OR70054-100G OR70054-25G OR70054-500G
APOSOR70054-100GEA 1240 CZK
APOSOR70054-100G APOSOR70054-25G APOSOR70054-500G
2-(tert-Butoxycarbonylamino)-1-ethanol
2-(tert-Butoxycarbonylamino)-1-ethanol
Vzorec: C₇H₁₅NO₃
MW: 161,20 g/mol
Bod varu: 92 °C/0,22mmHg
Hustota: 1,042 g/cm³ (25 °C)
Bod vzplanutí: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00056657
CAS číslo: 26690-80-2
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR