Tetrahydropyran-3-carbaldehyde
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR70049-1G
OR70049-2.5G
OR70049-250MG
APOSOR70049-1GEA
2860
CZK
APOSOR70049-1G
APOSOR70049-2.5G
APOSOR70049-250MG
Tetrahydropyran-3-carbaldehyde
Tetrahydropyran-3-carbaldehyde
Vzorec:
C₆H₁₀O₂ Bod varu: 54…55 °C/7mmHg Hustota: 1,0609 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD03789633 CAS číslo: 77342-93-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...