3-(Bromomethyl)tetrahydro-2H-pyran
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR70048-1G
OR70048-5G
APOSOR70048-1GEA
7440
CZK
APOSOR70048-1G
APOSOR70048-5G
3-(Bromomethyl)tetrahydro-2H-pyran
3-(Bromomethyl)tetrahydro-2H-pyran
Vzorec:
C₆H₁₁BrO MW: 179.06 g/mol Bod varu: 63…65 °C/5mmHg Hustota: 1.432 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD08235012 CAS číslo: 116131-44-3 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...