Tisk…

Ethyl-2-(5-chloro-1H-indazol-3-yl)acetate

Dodavatel: Apollo Scientific
Nebezpečí

OR70045-1G OR70045-250MG
APOSOR70045-1GEA 5290 CZK
APOSOR70045-1G APOSOR70045-250MG
Ethyl-2-(5-chloro-1H-indazol-3-yl)acetate
Ethyl-2-(5-chloro-1H-indazol-3-yl)acetate
Vzorec: C₁₁H₁₁ClN₂O₂
MW: 238,68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00056935
CAS číslo: 27512-72-7
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR