3-Bromo-5-methoxybenzyl alcohol
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(3-Bromo-5-methoxyphenyl)methanol
OR70038-1G
OR70038-250MG
OR70038-5G
APOSOR70038-1GEA
2940
CZK
APOSOR70038-1G
APOSOR70038-250MG
APOSOR70038-5G
3-Bromo-5-methoxybenzyl alcohol
3-Bromo-5-methoxybenzyl alcohol
Vzorec:
C₈H₉BrO₂ MW: 217,599 g/mol Bod tání: 45…46 °C Storage Temperature: Ambient |
MDL Number:
MFCD00229988 CAS číslo: 262450-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...