5-Bromo-2-pyridineacetic acid
Dodavatel: Apollo Scientific
Synonyms:
2-(5-Bromopyridin-2-yl)acetic acid
OR70028-1G
OR70028-5G
APOSOR70028-1GEA
5000
CZK
APOSOR70028-1G
APOSOR70028-5G
5-Bromo-2-pyridineacetic acid
5-Bromo-2-pyridineacetic acid
Vzorec:
C₇H₆BrNO₂ MW: 216,034 g/mol Bod tání: 125…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD09999983 CAS číslo: 192642-85-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...