2,6-Dimethylbenzylamine 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR70001-5G
OR70001-1G
APOSOR70001-5GEA
3640
CZK
APOSOR70001-5G
APOSOR70001-1G
2,6-Dimethylbenzylamine 98%
2,6-Dimethylbenzylamine
Vzorec:
C₉H₁₃N MW: 135,21 g/mol Bod varu: 111…114 °C/15mmHg Storage Temperature: Ambient |
CAS číslo:
74788-82-2 UN: 2735 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...