Tisk…

2-Hydroxy-p-toluic acid 99%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Hydroxy-4-methylbenzoic acid, m-Homosalicyclic acid, m-Cresotic acid, 4-Methylsalicylic acid

OR6995-100G OR6995-1KG OR6995-500G
APOSOR6995-100GEA 1040 CZK
APOSOR6995-100G APOSOR6995-1KG APOSOR6995-500G
2-Hydroxy-p-toluic acid 99%
2-Hydroxy-p-toluic acid
Vzorec: C₈H₈O₃
MW: 152.15
Bod tání: 173…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00002454
CAS číslo: 50-85-1
EINECS: 200-068-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR