Tisk…

5-Bromo-2-(methylthio)pyrimidine-4-carboxylic acid 95%

Dodavatel: Apollo Scientific
Varování

OR6988-1G OR6988-25G
APOSOR6988-1GEA 622 CZK
APOSOR6988-1G APOSOR6988-25G
5-Bromo-2-(methylthio)pyrimidine-4-carboxylic acid 95%
5-Bromo-2-(methylthio)pyrimidine-4-carboxylic acid
Vzorec: C₆H₅BrN₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD00238866
CAS číslo: 50593-92-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR