Tisk…

5-Nitro-2,4-xylidine

Dodavatel: Apollo Scientific
Varování

Synonyms: 2,4-Dimethyl-5-nitroaniline, 5-Nitro-2,4-xylidine

OR6966-1G OR6966-25G OR6966-5G
APOSOR6966-1GEA 622 CZK
APOSOR6966-1G APOSOR6966-25G APOSOR6966-5G
5-Nitro-2,4-xylidine
5-Nitro-2,4-xylidine
Vzorec: C₈H₁₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00025199
CAS číslo: 2124-47-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR