2-Chlorophenyl methyl sulphone 99%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1-Chloro-2-(methylsulphonyl)benzene
OR6950-1G
OR6950-25G
OR6950-5G
APOSOR6950-1GEA
622
CZK
APOSOR6950-1G
APOSOR6950-25G
APOSOR6950-5G
2-Chlorophenyl methyl sulphone 99%
2-Chlorophenyl methyl sulphone
Vzorec:
C₇H₇ClO₂S MW: 190,65 g/mol Bod tání: 91,5…92.5 °C Storage Temperature: Ambient |
CAS číslo:
17482-05-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...