Tisk…

2,2,6,6-Tetramethyl-4-piperidone hydrochloride 97%

Dodavatel: Apollo Scientific
Varování

OR6946-100G OR6946-250G OR6946-25G OR6946-50G
APOSOR6946-100GEA 6220 CZK
APOSOR6946-100G APOSOR6946-250G APOSOR6946-25G APOSOR6946-50G
2,2,6,6-Tetramethyl-4-piperidone hydrochloride 97%
2,2,6,6-Tetramethyl-4-piperidone hydrochloride
Vzorec: C₉H₁₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD00012768
CAS číslo: 33973-59-0
EINECS: 251-769-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR