Tisk…

Methyl-8-hydroxy-1-naphthoate

Dodavatel: Apollo Scientific
Varování

OR6939-250MG OR6939-100MG
APOSOR6939-250MGEA 2480 CZK
APOSOR6939-250MG APOSOR6939-100MG
Methyl-8-hydroxy-1-naphthoate
Methyl-8-hydroxy-1-naphthoate
Vzorec: C₁₂H₁₀O₃
MW: 202,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00665832
CAS číslo: 5247-86-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR