Tisk…

(±)-Pipecolinic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (±)-2-Piperidinecarboxylic acid, DL-Pipecolinic acid, H-DL-Homopro-OH, Pipecolinic Acid, Piperidine-2-carboxylic acid, Pipecolinic acid

OR6924-10G OR6924-25G
APOSOR6924-10GEA 622 CZK
APOSOR6924-10G APOSOR6924-25G
(±)-Pipecolinic acid 98%
(±)-Pipecolinic acid
Vzorec: C₆H₁₁NO₂
MW: 129.16
Bod tání: 280…283 °C
Storage Temperature: Ambient
MDL Number: MFCD00064347
CAS číslo: 535-75-1
EINECS: 223-737-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR