Tisk…

N,N-Diethyl-N-(3-piperazin-1-ylpropyl)amine 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-(3-Diethylaminopropyl)piperazine

OR6916-5G OR6916-1G
APOSOR6916-5GEA 5960 CZK
APOSOR6916-5G APOSOR6916-1G
N,N-Diethyl-N-(3-piperazin-1-ylpropyl)amine 98%
N,N-Diethyl-N-(3-piperazin-1-ylpropyl)amine
Vzorec: C₁₁H₂₅N₃
MW: 199,34 g/mol
Bod varu: 105…106 °C/3mmHg
Hustota: 0,9063 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01075182
CAS číslo: 22764-55-2
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR