1-[2-(Morpholin-4-yl)ethyl]piperazine 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-[2-(Piperazin-1-yl)ethyl]morpholine, 1-[2-(Morpholin-4-yl)-ethyl]-piperazine
OR6912-1G
OR6912-25G
OR6912-5G
APOSOR6912-1GEA
622
CZK
APOSOR6912-1G
APOSOR6912-25G
APOSOR6912-5G
1-[2-(Morpholin-4-yl)ethyl]piperazine 98%
1-[2-(Morpholin-4-yl)ethyl]piperazine
Vzorec:
C₁₀H₂₁N₃O MW: 199,298 g/mol Bod varu: 106…107 °C/0,2mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00191217 CAS číslo: 4892-89-1 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...