1-Isopropylpiperazine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR6901-5G
OR6901-1G
OR6901-25G
APOSOR6901-5GEA
705
CZK
APOSOR6901-5G
APOSOR6901-1G
APOSOR6901-25G
1-Isopropylpiperazine 97%
1-Isopropylpiperazine
Vzorec:
C₇H₁₆N₂ MW: 128.22 Bod varu: 180…181 °C/760mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00167971 CAS číslo: 4318-42-7 EINECS: 224-345-3 UN: 1992 ADR: 3+6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...