Tisk…

1-Butylpiperazine

Dodavatel: Apollo Scientific
Varování

OR6893-10G OR6893-1G OR6893-5G OR6893-25G
APOSOR6893-10GEA 2900 CZK
APOSOR6893-10G APOSOR6893-1G APOSOR6893-5G APOSOR6893-25G
1-Butylpiperazine
1-Butylpiperazine
Vzorec: C₈H₁₈N₂
MW: 142,24 g/mol
Bod varu: 116 °C/11mmHg
Storage Temperature: Ambient
MDL Number: MFCD00039702
CAS číslo: 5610-49-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR