Tisk…

1-[2-(1-Pyrrolidinyl)ethyl]piperazine 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR6890-25G OR6890-5G
APOSOR6890-25GEA 17430 CZK
APOSOR6890-25G APOSOR6890-5G
1-[2-(1-Pyrrolidinyl)ethyl]piperazine 97%
1-[2-(1-Pyrrolidinyl)ethyl]piperazine
Vzorec: C₁₀H₂₁N₃
MW: 183,30 g/mol
Bod varu: 77…79 °C
Storage Temperature: Ambient
MDL Number: MFCD01075190
CAS číslo: 22763-69-5
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR