Tisk…

N-Piperazineacetic acid morpholine amide 96%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(Morpholin-4-yl)-2-(piperazin-1-yl)ethan-1-one

OR6877-10G OR6877-1G
APOSOR6877-10GEA 5200 CZK
APOSOR6877-10G APOSOR6877-1G
N-Piperazineacetic acid morpholine amide 96%
N-Piperazineacetic acid morpholine amide
Vzorec: C₁₀H₁₉N₃O₂
MW: 213,28 g/mol
Bod tání: 63…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00006172
CAS číslo: 39890-46-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR