Cyclohexyltriphenylphosphonium bromide 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR6822-100G
OR6822-25G
OR6822-5G
APOSOR6822-100GEA
11410
CZK
APOSOR6822-100G
APOSOR6822-25G
APOSOR6822-5G
Cyclohexyltriphenylphosphonium bromide 97%
Cyclohexyltriphenylphosphonium bromide
Vzorec:
C₂₄H₂₆BrP Bod tání: 265…267 °C Storage Temperature: Ambient |
MDL Number:
MFCD00031667 CAS číslo: 7333-51-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...