(2,6-Dichlorophenyl)acetic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2,6-Dichlorophenylacetic acid
OR6814-100G
OR6814-25G
OR6814-10G
APOSOR6814-100GEA
2900
CZK
APOSOR6814-100G
APOSOR6814-25G
APOSOR6814-10G
(2,6-Dichlorophenyl)acetic acid 98%
(2,6-Dichlorophenyl)acetic acid
Vzorec:
(C₆H₃Cl₂)CH₂COOH MW: 205,04 g/mol Bod tání: 158…161 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004320 CAS číslo: 6575-24-2 EINECS: 229-504-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...