2-Propylvaleryl chloride 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Propylpentanoyl chloride, 2,2-Di-n-propylacetylchloride
OR6808-25G
OR6808-5G
APOSOR6808-25GEA
9430
CZK
APOSOR6808-25G
APOSOR6808-5G
2-Propylvaleryl chloride 98%
2-Propylvaleryl chloride
Vzorec:
C₈H₁₅ClO MW: 162,66 g/mol Bod varu: 68…70 °C/10mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00051446 CAS číslo: 2936-08-5 UN: 3265 ADR: 8,II |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...