Tisk…

Methyl-4-(aminomethyl)benzoate hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-(Aminomethyl)benzoic Acid Methyl Ester Hydrochloride, Methyl 4-(aminomethyl)benzoate hydrochloride

OR6787-25G OR6787-5G
APOSOR6787-25GEA 2280 CZK
APOSOR6787-25G APOSOR6787-5G
Methyl-4-(aminomethyl)benzoate hydrochloride
Methyl-4-(aminomethyl)benzoate hydrochloride
Vzorec: C₉H₁₁NO₂
MW: 201,65 g/mol
Bod tání: 234…243 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00182671
CAS číslo: 6232-11-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR