4-Biphenylmethyl chloride
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-(Chloromethyl)biphenyl
OR6777-1G
OR6777-25G
OR6777-5G
APOSOR6777-1GEA
622
CZK
APOSOR6777-1G
APOSOR6777-25G
APOSOR6777-5G
4-Biphenylmethyl chloride
4-Biphenylmethyl chloride
Vzorec:
C₁₃H₁₁Cl MW: 202.69 g/mol Bod tání: 71…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000917 CAS číslo: 1667-11-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...