Tisk…

Ethyl 5-(Thiophen-2-yl)isoxazole-3-carboxylate 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl 5-(thiophen-2-yl)isoxazole-3-carboxylate, 5-(Thiophen-2-yl)isoxazole-3-carboxylic acid Ethyl ester

OR6758-1G OR6758-5G
APOSOR6758-1GEA 4770 CZK
APOSOR6758-1G APOSOR6758-5G
Ethyl 5-(Thiophen-2-yl)isoxazole-3-carboxylate 97%
Ethyl 5-(Thiophen-2-yl)isoxazole-3-carboxylate
Vzorec: C₁₀H₉NO₃S
MW: 223,25 g/mol
Bod tání: 48…51 °C
Storage Temperature: Ambient
MDL Number: MFCD02090509
CAS číslo: 90924-54-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR