4-(Piperidin-1-ylmethyl)benzaldehyde 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR6740-1G
OR6740-250MG
OR6740-5G
APOSOR6740-1GEA
6220
CZK
APOSOR6740-1G
APOSOR6740-250MG
APOSOR6740-5G
4-(Piperidin-1-ylmethyl)benzaldehyde 95%
4-(Piperidin-1-ylmethyl)benzaldehyde
Vzorec:
C₁₃H₁₇NO Storage Temperature: Ambient |
MDL Number:
MFCD07772862 CAS číslo: 471929-86-9 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...