(±)-β-Methylphenethylamine 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Phenylpropylamine
OR6734-1G
OR6734-25G
OR6734-5G
APOSOR6734-1GEA
2200
CZK
APOSOR6734-1G
APOSOR6734-25G
APOSOR6734-5G
(±)-β-Methylphenethylamine 98%
(±)-β-Methylphenethylamine
Vzorec:
C₉H₁₃N Bod varu: 82…84 °C/10mmHg Hustota: 0,93 g/cm³ (25 °C) Bod vzplanutí: 79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008144 CAS číslo: 582-22-9 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...