Tisk…

(4-(Pyrrolidin-1-yl)phenyl)methanamine 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-(Pyrrolidin-1-yl)benzylamine

OR6730-1G OR6730-250MG OR6730-5G
APOSOR6730-1GEA 3110 CZK
APOSOR6730-1G APOSOR6730-250MG APOSOR6730-5G
(4-(Pyrrolidin-1-yl)phenyl)methanamine 97%
(4-(Pyrrolidin-1-yl)phenyl)methanamine
Vzorec: C₁₁H₁₆N₂
MW: 176,2581 g/mol
Bod tání: 47…50 °C
Storage Temperature: Ambient
MDL Number: MFCD04971090
CAS číslo: 114365-04-7
UN: 3259
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR