Tisk…

2,6-Dichloro-3-iodopyridine 95%

Dodavatel: Apollo Scientific
Nebezpečí

OR6713-10G
APOSOR6713-10GEA 500 CZK
APOSOR6713-10G
2,6-Dichloro-3-iodopyridine 95%
2,6-Dichloro-3-iodopyridine
Vzorec: C₅H₂Cl₂IN
MW: 273,88 g/mol
Bod tání: 74…79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094686
CAS číslo: 148493-37-2
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR