4-Benzyloxyaniline 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-(Benzyloxy)aniline
OR6680-100G
OR6680-25G
APOSOR6680-100GEA
5530
CZK
APOSOR6680-100G
APOSOR6680-25G
4-Benzyloxyaniline 98%
4-Benzyloxyaniline
Vzorec:
C₁₃H₁₃NO MW: 199,2484 g/mol Bod tání: 45…46 °C Storage Temperature: Ambient |
MDL Number:
MFCD00025318 CAS číslo: 6373-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...