Tisk…

4-Methylsulphonylbenzylamine hydrochloride 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: (4-(Methylsulphonyl)phenyl)methanamine hydrochloride

OR6633-25G OR6633-5G
APOSOR6633-25GEA 21330 CZK
APOSOR6633-25G APOSOR6633-5G
4-Methylsulphonylbenzylamine hydrochloride 95%
4-Methylsulphonylbenzylamine hydrochloride
Vzorec: C₈H₁₁NO₂S.HCl
MW: 221,70 g/mol
Bod tání: 272…274 °C
Storage Temperature: Ambient
MDL Number: MFCD00216491
CAS číslo: 98593-51-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR