Tisk…

4-Amino-m-anisic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Amino-3-methoxybenzenecarboxylic acid

OR6621-100G OR6621-25G OR6621-5G
APOSOR6621-100GEA 8670 CZK
APOSOR6621-100G APOSOR6621-25G APOSOR6621-5G
4-Amino-m-anisic acid
4-Amino-m-anisic acid
Vzorec: C₈H₉NO₃
Bod tání: 186…188 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00016539
CAS číslo: 2486-69-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR