3-Bromo-5-iodopyridine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR6617-1G
OR6617-250MG
OR6617-5G
APOSOR6617-1GEA
705
CZK
APOSOR6617-1G
APOSOR6617-250MG
APOSOR6617-5G
3-Bromo-5-iodopyridine
3-Bromo-5-iodopyridine
Vzorec:
C₅H₃BrIN MW: 283.89 Bod tání: 127…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD03086019 CAS číslo: 233770-01-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...