Methyl-2-naphthoate 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 2-naphthoate
OR6584-100G
OR6584-25G
OR6584-5G
APOSOR6584-100GEA
17860
CZK
APOSOR6584-100G
APOSOR6584-25G
APOSOR6584-5G
Methyl-2-naphthoate 97%
Methyl-2-naphthoate
Vzorec:
C₁₂H₁₀O₂ MW: 186,21 g/mol Bod varu: 141…143 °C/4mmHg Bod tání: 71…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017345 CAS číslo: 2459-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...