2-Nitrocinnamic acid 99%
Dodavatel: Apollo Scientific
OR6581-100G
OR6581-25G
OR6581-500G
APOSOR6581-100GEA
8960
CZK
APOSOR6581-100G
APOSOR6581-25G
APOSOR6581-500G
2-Nitrocinnamic acid 99%
2-Nitrocinnamic acid
Vzorec:
C₉H₇NO₄ Bod tání: 243…245 °C Storage Temperature: Ambient |
CAS číslo:
612-41-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...