4-Bromocinnamic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-(4-Bromophenyl)acrylic acid
OR6580-100G
OR6580-500G
OR6580-1KG
APOSOR6580-100GEA
3740
CZK
APOSOR6580-100G
APOSOR6580-500G
APOSOR6580-1KG
4-Bromocinnamic acid 98%
4-Bromocinnamic acid
Vzorec:
C₉H₇BrO₂ MW: 227.06 Bod tání: 260…264 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004394 CAS číslo: 1200-07-3 EINECS: 214-850-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...