Thioxanthen-9-one 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Thioxanthone
OR6559-100G
OR6559-25G
APOSOR6559-100GEA
5590
CZK
APOSOR6559-100G
APOSOR6559-25G
Thioxanthen-9-one 95%
Thioxanthen-9-one
Vzorec:
C₁₃H₈OS MW: 212,2700 g/mol Bod varu: 371…373 °C/715mmHg Bod tání: 210…213 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005066 CAS číslo: 492-22-8 EINECS: 207-749-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...