Benzimidazole 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1H-Benzo[d]imidazole, 1H-Benzimidazole
OR6549-50G
OR6549-100G
OR6549-500G
APOSOR6549-50GEA
622
CZK
APOSOR6549-50G
APOSOR6549-100G
APOSOR6549-500G
Benzimidazole 97%
Benzimidazole
Vzorec:
C₇H₆N₂ MW: 118,14 g/mol Bod varu: 360 °C Bod tání: 169…174 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005585 CAS číslo: 51-17-2 EINECS: 200-081-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...