Benzofuran 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2,3-Benzofuran, Benzo[b]furan, Benzo(b)furan, 1-Benzofuran, Coumarone
OR6548-100G
OR6548-25G
APOSOR6548-100GEA
2110
CZK
APOSOR6548-100G
APOSOR6548-25G
Benzofuran 98%
Benzofuran
Vzorec:
C₈H₆O MW: 118,14 g/mol Bod varu: 173…175 °C Hustota: 1,072 g/cm³ (25 °C) Bod vzplanutí: 56 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005847 CAS číslo: 271-89-6 EINECS: 205-982-6 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...