Tisk…

2-Aminoacetophenone hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: Phenacylamine hydrochloride

OR6519-100G OR6519-10G OR6519-25G
APOSOR6519-100GEA 7280 CZK
APOSOR6519-100G APOSOR6519-10G APOSOR6519-25G
2-Aminoacetophenone hydrochloride
2-Aminoacetophenone hydrochloride
Vzorec: C₈H₁₀ClNO
MW: 171,63 g/mol
Bod tání: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00012873
CAS číslo: 5468-37-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR