7-Bromoheptanoic acid 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
7-Bromoenanthic Acid
OR6483-5G
OR6483-10G
OR6483-1G
APOSOR6483-5GEA
910
CZK
APOSOR6483-5G
APOSOR6483-10G
APOSOR6483-1G
7-Bromoheptanoic acid 97%
7-Bromoheptanoic acid
Radiopharmaceutical precursor.
Vzorec:
C₇H₁₃BrO₂ MW: 209,08 g/mol Bod varu: 145 °C/3mmHg Bod tání: 27…32 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037800 CAS číslo: 30515-28-7 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...