Tisk…

Iodoantipyrine 98%

Dodavatel: Apollo Scientific

Synonyms: 4-Iodo-1,5-dimethyl-2-phenyl-1H-pyrazol-3(2H)-one

OR6470-100G OR6470-25G
APOSOR6470-100GEA 45300 CZK
APOSOR6470-100G APOSOR6470-25G
Iodoantipyrine 98%
Iodoantipyrine

Radiopharmaceutical precursor.

Vzorec: C₁₁H₁₁IN₂O
MW: 314,13 g/mol
Bod tání: 161…163 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00040720
CAS číslo: 129-81-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR