2-Chloroethyl isocyanate 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Isocyanic acid 2-Chloroethyl ester
OR6436-100G
APOSOR6436-100GEA
3970
CZK
APOSOR6436-100G
2-Chloroethyl isocyanate 97%
2-Chloroethyl isocyanate
- May form small amount of sediment.
Vzorec:
C₃H₄ClNO MW: 105,52 g/mol Bod varu: 135…142 °C Hustota: 1,237 g/cm³ (25 °C) Bod vzplanutí: 56 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00002043 CAS číslo: 1943-83-5 EINECS: 217-734-4 UN: 2478 ADR: 3+6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...